EMAIL THIS PAGE TO A FRIEND
02680 Sigma-Aldrich

Ethoxyacetic acid

purum, ≥98.0% (GC)

Synonym: O-Ethylglycolic acid

  • CAS Number 627-03-2

  • Linear Formula C2H5OCH2COOH

  • Molecular Weight 104.10

  •  Beilstein Registry Number 1743037

  •  EC Number 210-978-2

  •  MDL number MFCD00004310

  •  PubChem Substance ID 329747490

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   YZGQDNOIGFBYKF-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.419(lit.)
  n20/D 1.419
bp   97-100 °C/11 mmHg(lit.)
density   1.102 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
RTECS 
AH3600000
Flash Point(F) 
206.6 °F
Flash Point(C) 
97 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

137111 Ethoxyacetic acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?