• USA Home
  • 02681 - Gold atomic spectroscopy standard concentrate 1.00 g Au

EMAIL THIS PAGE TO A FRIEND
02681 Sigma-Aldrich

Gold atomic spectroscopy standard concentrate 1.00 g Au

1.00 g/L, for 1L standard solution, analytical standard

Synonym: Gold(III) chloride solution

Purchase

Properties

Related Categories 1.00 g/L, AAS, AAS Concentrates, Additional Standards, Analytical Standards,
grade   analytical standard
InChI Key   PCHJSUWPFVWCPO-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
packaging   ampule of
solubility   H2O: soluble

Description

Packaging

glass ampoule

Preparation Note

prepared with HAuCl4, HCl and H2O

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1789 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

94791 SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules
43917 SnapIT Ampule Opener, large, for 5-10ml, 10-15ml, 20-25ml ampoules

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?