• USA Home
  • 02685 - Lithium atomic spectroscopy standard concentrate 1.00 g Li

EMAIL THIS PAGE TO A FRIEND
02685 Sigma-Aldrich

Lithium atomic spectroscopy standard concentrate 1.00 g Li

for 1 l standard solution, 1.00 g/L, analytical standard

Synonym: Lithium chloride solution

Purchase

Properties

Related Categories 1.00 g/L, AAS, AAS Concentrates, Additional Standards, Analytical Standards,
grade   analytical standard
InChI Key   KWGKDLIKAYFUFQ-UHFFFAOYSA-M
shelf life   limited shelf life, expiry date on the label
packaging   ampule of
concentration   1.00 g/L
solubility   H2O: soluble

Description

Preparation Note

prepared with LiCl and H2O

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
OJ5950000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?