• USA Home
  • 02688 - Silver atomic spectroscopy standard concentrate 1.00 g Ag

EMAIL THIS PAGE TO A FRIEND
02688 Sigma-Aldrich

Silver atomic spectroscopy standard concentrate 1.00 g Ag

1.00 g/L, for 1L standard solution

Synonym: Silver nitrate solution

  • CAS Number 7761-88-8

  • Empirical Formula (Hill Notation) AgNO3

  • Molecular Weight 169.87

  •  E Number E174

  •  MDL number MFCD00003414

  •  PubChem Substance ID 329747497

Purchase

Properties

Related Categories AAS, Additional Standards, Analytical Standards, Analytical/Chromatography, Single Solution,
InChI Key   SQGYOTSLMSWVJD-UHFFFAOYSA-N
quality   ampule
shelf life   limited shelf life, expiry date on the label
concentration   1.00 g/L
solubility   H2O: soluble

Description

Preparation Note

prepared with AgNO3, HNO3 and H2O

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3264 8/PG 2
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?