EMAIL THIS PAGE TO A FRIEND
02690 Sigma-Aldrich

Ethyl ethoxyacetate

purum, ≥97.5% (GC)

Synonym: Diethyl glycolate

  • CAS Number 817-95-8

  • Linear Formula CH3CH2OCH2COOCH2CH3

  • Molecular Weight 132.16

  •  Beilstein Registry Number 1749545

  •  EC Number 212-447-0

  •  MDL number MFCD00040492

  •  PubChem Substance ID 329747498

Purchase

Properties

Related Categories Building Blocks, C6 to C7, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   CKSRFHWWBKRUKA-UHFFFAOYSA-N
grade   purum
assay   ≥97.5% (GC)
refractive index   n20/D 1.402(lit.)
  n20/D 1.403
bp   156 °C(lit.)
density   0.975 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3272 3 / PGIII
WGK Germany 
3
Flash Point(F) 
122 °F
Flash Point(C) 
50 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

374172 Ethyl ethoxyacetate, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?