• USA Home
  • 02740 - Ethyl 2-cyano-3-ethoxyacrylate

EMAIL THIS PAGE TO A FRIEND
02740 Sigma-Aldrich

Ethyl 2-cyano-3-ethoxyacrylate

purum, ≥97.0% (GC)

Synonym: Ethyl (ethoxymethylene)cyanoacetate

  • CAS Number 94-05-3

  • Linear Formula C2H5OCH=C(CN)COOC2H5

  • Molecular Weight 169.18

  •  Beilstein Registry Number 878814

  •  EC Number 202-299-5

  •  MDL number MFCD00009136

  •  PubChem Substance ID 24845376

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   KTMGNAIGXYODKQ-VOTSOKGWSA-N
grade   purum
assay   ≥97.0% (GC)
bp   190-191 °C/30 mmHg(lit.)
mp   49-51 °C(lit.)
  49-52 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
1
Flash Point(F) 
266 °F
Flash Point(C) 
130 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?