• USA Home
  • 02762 - Tin atomic spectroscopy standard concentrate 1.00 g Sn

EMAIL THIS PAGE TO A FRIEND
02762 Sigma-Aldrich

Tin atomic spectroscopy standard concentrate 1.00 g Sn

1.00 g/L, for 1L standard solution, analytical standard

Synonym: Tin(II) chloride solution

Purchase

Properties

Related Categories 1.00 g/L, AAS, AAS Concentrates, Additional Standards, Analytical Standards,
grade   analytical standard
InChI Key   ATJFFYVFTNAWJD-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
packaging   ampule of
solubility   H2O: soluble

Description

Preparation Note

prepared with SnCl2.2H2O, HCl and H2O

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN3264 - class 8 - PG 2 - EHS - Corrosive liquid, acidic, inorganic, n.o.s., HI: all
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?