• USA Home
  • 02781 - Aluminum atomic spectroscopy standard concentrate 1.00 g Al

EMAIL THIS PAGE TO A FRIEND
02781 Sigma-Aldrich

Aluminum atomic spectroscopy standard concentrate 1.00 g Al

1.00 g/L, for 1 l standard solution, analytical standard

Synonym: Aluminum chloride solution

Purchase

Properties

Related Categories 1.00 g/L, AAS, AAS Concentrates, Additional Standards, Analytical Standards,
grade   analytical standard
InChI Key   VSCWAEJMTAWNJL-UHFFFAOYSA-K
shelf life   limited shelf life, expiry date on the label
packaging   ampule of
concentration   1.00 g/L
solubility   H2O: soluble

Description

Preparation Note

prepared with AlCl3.6H2O and H2O

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 2581 8 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?