• USA Home
  • 02784 - Barium atomic spectroscopy standard concentrate 1.00 g Ba

EMAIL THIS PAGE TO A FRIEND
02784 Sigma-Aldrich

Barium atomic spectroscopy standard concentrate 1.00 g Ba

1.00 g/L, for 1L standard solution

Synonym: Barium chloride solution

  • CAS Number 10361-37-2

  • Empirical Formula (Hill Notation) BaCl2

  • Molecular Weight 208.23

  •  MDL number MFCD00003445

Purchase

Properties

Related Categories 1.00 g/L, AAS, AAS Concentrates, Additional Standards, Analytical Standards,
InChI Key   WDIHJSXYQDMJHN-UHFFFAOYSA-L
quality   ampule
shelf life   limited shelf life, expiry date on the label
solubility   H2O: soluble

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Preparation Note

prepared with BaCl2.2H2O and H2O

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?