• USA Home
  • 02787 - Cesium atomic spectroscopy standard concentrate 1.00 g Cs

EMAIL THIS PAGE TO A FRIEND
02787 Sigma-Aldrich

Cesium atomic spectroscopy standard concentrate 1.00 g Cs

1.00 g/L, for 1 l standard solution

Synonym: Cesium chloride solution

Purchase

Properties

Related Categories 1.00 g/L, AAS, AAS Concentrates, Additional Standards, Analytical Standards,
InChI Key   TVFDJXOCXUVLDH-UHFFFAOYSA-N
quality   ampule
shelf life   limited shelf life, expiry date on the label
concentration   1.00 g/L
solubility   H2O: soluble

Description

Preparation Note

prepared with CsCl and H2O

Price and Availability

Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?