• USA Home
  • 02794 - Multielement Cd, Cr, Cu, Fe, Mn, Ni, Pb, Sb, Sn, Zn atomic spectroscopy standard concentrate

EMAIL THIS PAGE TO A FRIEND
02794 Sigma-Aldrich

Multielement Cd, Cr, Cu, Fe, Mn, Ni, Pb, Sb, Sn, Zn atomic spectroscopy standard concentrate

for ICP

Purchase

Properties

Related Categories AAS, Additional Standards, Analytical Standards, Analytical/Chromatography, Elemental & Metal Standards,
grade   analytical standard
  for ICP
shelf life   limited shelf life, expiry date on the label

Description

Components

Cd, Cr, Cu, Fe, Mn, Ni, Pb, Sb, Sn, Zn each with 1 mg in diluted hydrochloric acid

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN3264 - class 8 - PG 2 - EHS - Corrosive liquid, acidic, inorganic, n.o.s., HI: all
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?