EMAIL THIS PAGE TO A FRIEND
02900 Sigma-Aldrich

Allyl ethyl ether

technical, ≥95% (GC)

Synonym: 3-Ethoxypropene, Ethyl allyl ether

  • CAS Number 557-31-3

  • Linear Formula C2H5OCH2CH=CH2

  • Molecular Weight 86.13

  •  Beilstein Registry Number 1361480

  •  EC Number 209-169-7

  •  MDL number MFCD00009255

  •  PubChem Substance ID 24845409

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
InChI Key   OJPSFJLSZZTSDF-UHFFFAOYSA-N
grade   technical
assay   ≥95% (GC)
refractive index   n20/D 1.388(lit.)
  n20/D 1.388
bp   65-66 °C(lit.)
density   0.76 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2335 6.1(3) / PGII
WGK Germany 
3
Flash Point(F) 
-5.8 °F
Flash Point(C) 
-21 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?