EMAIL THIS PAGE TO A FRIEND
02950 Sigma-Aldrich

Ethylamine solution

purum, 70% in H2O

Synonym: Aminoethane, Monoethylamine

  • CAS Number 75-04-7

  • Linear Formula C2H5NH2

  • Molecular Weight 45.08

  •  Beilstein Registry Number 505933

  •  MDL number MFCD00008160

Purchase

Properties

Related Categories Acids & Bases, Bases, Chemical Synthesis, Organic Bases, Synthetic Reagents More...
InChI Key   QUSNBJAOOMFDIB-UHFFFAOYSA-N
grade   purum
concentration   70% in H2O
refractive index   n20/D 1.384
density   0.81 g/mL at 20 °C

Description

Other Notes

Sales restrictions may apply

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2270 8(3) / PGII
WGK Germany 
1
Flash Point(F) 
<-11.2 °F
Flash Point(C) 
<-24 °C

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?