• USA Home
  • 03125 - 1-Bromo-2-ethylbenzene

EMAIL THIS PAGE TO A FRIEND
03125 Sigma-Aldrich

1-Bromo-2-ethylbenzene

puriss., ≥98.5% (GC)

  • CAS Number 1973-22-4

  • Linear Formula C2H5C6H4Br

  • Molecular Weight 185.06

  •  Beilstein Registry Number 1929735

  •  EC Number 217-823-8

  •  MDL number MFCD00000077

Purchase

Properties

Related Categories Aryl, Building Blocks, C8, Chemical Synthesis, Halogenated Hydrocarbons,
InChI Key   HVRUGFJYCAFAAN-UHFFFAOYSA-N
grade   puriss.
assay   ≥98.5% (GC)
refractive index   n20/D 1.549(lit.)
  n20/D 1.550
bp   199 °C(lit.)
density   1.338 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN3082 - class 9 - PG 3 - DOT NA1993 - Environmentally hazardous substances, liquid, n.o.s. HI: all (not BR)
WGK Germany 
3
Flash Point(F) 
170.6 °F
Flash Point(C) 
77 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

343315 1-Bromo-2-ethylbenzene, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?