• USA Home
  • 03261 - Cellulose acetate sheets, humide

EMAIL THIS PAGE TO A FRIEND
03261 Sigma-Aldrich

Cellulose acetate sheets, humide

size 7.8 cm × 15.0 cm

Purchase

Properties

Related Categories Cellulose Electrophoresis, Molecular Biology, Protein Electrophoresis, Proteomics
size   7.8 cm × 15.0 cm

Description

Application

For use in cellulose acetate electrophoresis

General description

After opening the package store membranes in 30% methanol. Do not let the membrane become dry. Before use store membranes for 10 minutes minimum in buffer. Apply sample on dull side. (Cut corner is bottom right!). Storage solution contains methanol.

After opening, store in 30% methanol. Do not let the membrane become dry. Before use, equilibrate membranes for at least 10 minutes in buffer. Apply sample on dull side (with the cut corner at bottom right). Storage solution contains methanol.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3175 4.1 / PGII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?