• USA Home
  • 03364 - Sulfate atomic spectroscopy standard concentrate 10.00 g SO42-

EMAIL THIS PAGE TO A FRIEND
03364 Sigma-Aldrich

Sulfate atomic spectroscopy standard concentrate 10.00 g SO42-

10.00 g/L, for 1 l standard solution, analytical standard

Synonym: Sulfuric acid standard solution

Purchase

Properties

Related Categories 10.00 g/L, AAS, AAS Concentrates, Additional Standards, Analytical Standards,
grade   analytical standard
InChI Key   QAOWNCQODCNURD-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
packaging   ampule of
concentration   10.00 g/L
solubility   H2O: soluble

Description

Preparation Note

prepared with H2SO4 and H2O

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?