• USA Home
  • 03366 - Phosphate atomic spectroscopy standard concentrate 1.00 g PO43-

EMAIL THIS PAGE TO A FRIEND
03366 Sigma-Aldrich

Phosphate atomic spectroscopy standard concentrate 1.00 g PO43-

1.00 g/L, for 1L standard solution, analytical standard

Synonym: Phosphoric acid solution

  • CAS Number 7664-38-2

  • Linear Formula H3PO4

  • Molecular Weight 98.00

  •  Beilstein Registry Number 1921286

  •  MDL number MFCD00011340

  •  PubChem Substance ID 329747603

Purchase

Properties

Related Categories 1.00 g/L, AAS, AAS Concentrates, Additional Standards, Analytical Standards,
grade   analytical standard
InChI Key   NBIIXXVUZAFLBC-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
packaging   ampule of
concentration   1.00 g/L
solubility   H2O: soluble
Gene Information   human ... SRC(6714)

Description

Preparation Note

prepared with H3PO4 and H2O

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?