• USA Home
  • 03367 - Phosphate atomic spectroscopy standard concentrate 10.00 g PO43-

EMAIL THIS PAGE TO A FRIEND
03367 Sigma-Aldrich

Phosphate atomic spectroscopy standard concentrate 10.00 g PO43-

10.00 g/L, for 1 l standard solution, analytical standard

Synonym: Phosphoric acid solution

  • CAS Number 7664-38-2

  • Linear Formula H3PO4

  • Molecular Weight 98.00

  •  Beilstein Registry Number 1921286

  •  MDL number MFCD00011340

  •  PubChem Substance ID 24845459

Purchase

Properties

Related Categories 10.00 g/L, AAS, AAS Concentrates, Additional Standards, Analytical Standards,
grade   analytical standard
InChI Key   NBIIXXVUZAFLBC-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
packaging   ampule of
concentration   10.00 g/L
solubility   H2O: soluble
Gene Information   human ... SRC(6714)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Preparation Note

prepared with H3PO4 and H2O

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?