• USA Home
  • 03370 - Sodium atomic spectroscopy standard concentrate 10.00 g Na

EMAIL THIS PAGE TO A FRIEND
03370 Sigma-Aldrich

Sodium atomic spectroscopy standard concentrate 10.00 g Na

10.00 g/L, for 1 l standard solution, analytical standard

Synonym: Sodium chloride solution

  • CAS Number 7647-14-5

  • Linear Formula NaCl

  • Molecular Weight 58.44

  •  Beilstein Registry Number 3534976

  •  MDL number MFCD00003477

  •  PubChem Substance ID 329747607

Purchase

Properties

Related Categories 10.00 g/L, AAS, AAS Concentrates, Additional Standards, Analytical Standards,
grade   analytical standard
InChI Key   FAPWRFPIFSIZLT-UHFFFAOYSA-M
shelf life   limited shelf life, expiry date on the label
packaging   ampule of
concentration   10.00 g/L
solubility   H2O: soluble

Description

Preparation Note

prepared with NaCl and H2O

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?