• USA Home
  • 03371 - Magnesium atomic spectroscopy standard concentrate 10.00 g Mg

EMAIL THIS PAGE TO A FRIEND
03371 Sigma-Aldrich

Magnesium atomic spectroscopy standard concentrate 10.00 g Mg

10.00 g/L, for 1 l standard solution, analytical standard

Synonym: Magnesium chloride solution

Purchase

Properties

Related Categories 10.00 g/L, AAS, AAS Concentrates, Additional Standards, Analytical Standards,
grade   analytical standard
InChI Key   TWRXJAOTZQYOKJ-UHFFFAOYSA-L
shelf life   limited shelf life, expiry date on the label
packaging   ampule of
concentration   10.00 g/L
solubility   H2O: soluble

Description

Preparation Note

prepared with MgCl2.6H2O and H2O

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?