EMAIL THIS PAGE TO A FRIEND
03420 Sigma-Aldrich

Dichloroethylsilane

≥98.0% (AT)

Synonym: Ethyldichlorosilane

  • CAS Number 1789-58-8

  • Empirical Formula (Hill Notation) C2H6Cl2Si

  • Molecular Weight 129.06

  •  Beilstein Registry Number 1731680

  •  EC Number 217-255-0

  •  MDL number MFCD00053205

  •  PubChem Substance ID 329747629

Purchase

Properties

Related Categories Chemical Synthesis, Protecting and Derivatizing Reagents, Protection and Derivatization, Silicon-Based, Synthetic Reagents More...
InChI Key   RKHXNBJRDQOIOQ-UHFFFAOYSA-N
assay   ≥98.0% (AT)
refractive index   n20/D 1.415
density   1.089 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1183 - class 4.3 - PG 1 - Ethyldichlorosilane
WGK Germany 
1
Flash Point(F) 
15.8 °F
Flash Point(C) 
-9 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?