• USA Home
  • 03442 - N,N-Dimethylethylamine

EMAIL THIS PAGE TO A FRIEND
03442 Sigma-Aldrich

N,N-Dimethylethylamine

purum, ≥99.0% (GC)

Synonym: N-Ethyldimethylamine, DMEA

Purchase

Properties

vapor pressure   8.09 psi ( 20 °C)
InChI Key   DAZXVJBJRMWXJP-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.372(lit.)
  n20/D 1.374
bp   36-38 °C(lit.)
mp   −140 °C(lit.)
density   0.675 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2733PSN1 8(3) / PGII
WGK Germany 
3
Flash Point(F) 
-18.4 °F
Flash Point(C) 
-28 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?