EMAIL THIS PAGE TO A FRIEND
03484 Sigma-Aldrich

Ethylene

≥99.9%

Synonym: Ethene

  • CAS Number 74-85-1

  • Linear Formula CH2=CH2

  • Molecular Weight 28.05

  •  Beilstein Registry Number 1730731

  •  EC Number 200-815-3

  •  MDL number MFCD00008604

Purchase

Properties

Related Categories Chemical Synthesis, Specialty Gases, Synthetic Reagents More...
vapor density   0.97 (vs air)
vapor pressure   35.04 atm ( 20 °C)
InChI Key   VGGSQFUCUMXWEO-UHFFFAOYSA-N
assay   ≥99.9%
autoignition temp.   842 °F
expl. lim.   36 %
impurities   ≤2000 ppm carbohydrates
bp   −104 °C(lit.)
mp   −169 °C(lit.)

Description

Other Notes

Sales restrictions may apply

Same product — same quality — now Sigma-Aldrich brand

Packaging

Cylinder with net ~3.2 kg

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1962 2.1
WGK Germany 
nwg
RTECS 
KU5340000
Flash Point(F) 
-148 °F
Flash Point(C) 
-100 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

control valve

Product #

Description

Add to Cart

99112 Outlet valve No 70688, stainless steel, pkg of 1 ea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?