• USA Home
  • 03583 - 1-(Boc-amino)cyclopentanecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
03583 Sigma-Aldrich

1-(Boc-amino)cyclopentanecarboxylic acid

≥98.0% (TLC)

Synonym: Boc-cycloleucine

  • CAS Number 35264-09-6

  • Linear Formula (CH3)3CO2CNHC5H8CO2H

  • Molecular Weight 229.27

  •  Beilstein Registry Number 2734412

  •  MDL number MFCD01076126

  •  PubChem Substance ID 329747736

Purchase

Properties

Related Categories Alicyclic Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   YBZCSKVLXBOFSL-UHFFFAOYSA-N
assay   ≥98.0% (TLC)

Description

Other Notes

This bulky cycloleucine imparts protection against hydrolysis to neighbouring amide bonds. Synthesis of conformationally restricted analogs.

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?