• USA Home
  • 03592 - Dimethyl N,N-diisopropylphosphoramidite

EMAIL THIS PAGE TO A FRIEND
03592 Sigma-Aldrich

Dimethyl N,N-diisopropylphosphoramidite

≥95.0% (GC)

  • CAS Number 122194-07-4

  • Empirical Formula (Hill Notation) C8H20NO2P

  • Molecular Weight 193.22

  •  Beilstein Registry Number 4242254

  •  MDL number MFCD00153505

  •  PubChem Substance ID 329747740

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Phosphoramidites, Phosphorus Compounds More...
InChI Key   KXUMNSXPAYCKPR-UHFFFAOYSA-N
assay   ≥95.0% (GC)
refractive index   n20/D 1.420(lit.)
bp   54 °C/10 mmHg(lit.)
density   0.840 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Reagent for oligonucleotide synthesis: synthesis of phosphoramidite derivatives

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?