• USA Home
  • 03593 - Viscosity validation set poly(methyl methacrylate) Mp 8′000-1′000′000 in THF

EMAIL THIS PAGE TO A FRIEND
03593 Sigma-Aldrich

Viscosity validation set poly(methyl methacrylate) Mp 8′000-1′000′000 in THF

certified according to DIN

Synonym: Viscosity validation set poly(methyl methacrylate) Mp 8′000-1′000′000 in THF certified according to DIN

Purchase

Description

Components

set contains 4 different poly(methyl methacrylate) standards with 500 mg with Mp ~8200, ~93000, ~175000, ~903000 for viscosimeters; each standard is characterized with GPC and viscosimetry

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?