• USA Home
  • 03594 - LALLS viscosity validation set polystyrene Mp 9′000-800′000 in TCB certified according to DIN

EMAIL THIS PAGE TO A FRIEND
03594 Sigma-Aldrich

LALLS viscosity validation set polystyrene Mp 9′000-800′000 in TCB certified according to DIN

certified reference material

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Application CRMs, Certified Reference Materials (CRMs),
grade   certified reference material
CofA   exact values can be taken from the accompanying certificate of analysis

Description

Components

set contains 4 different polystyrene standards with 500 mg with Mp ~9130, ~133000, ~221000, ~827000 for LALLS and viscosimeters; each standard is characterized with GPC, light scattering and viscosimetry

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?