• USA Home
  • 03645 - Ethylenediaminetetraacetic acid dilithium salt hydrate

EMAIL THIS PAGE TO A FRIEND
03645 Sigma-Aldrich

Ethylenediaminetetraacetic acid dilithium salt hydrate

≥97.0% (KT)

Synonym: Dilithium ethylenediaminetetraacetate hydrate

  • CAS Number 14531-56-7

  • Linear Formula (HOOCCH2)2NCH2CH2N(CH2COOLi)2 · aq

  • Molecular Weight 304.11 (anhydrous basis)

  •  EC Number 238-557-9

  •  MDL number MFCD00150462

  •  PubChem Substance ID 24845524

Purchase

Properties

InChI Key   KRGLZFVNTDZMFB-UHFFFAOYSA-L
assay   ≥97.0% (KT)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?