• USA Home
  • 03696 - Fmoc-β-Homoser(tBu)-OH

EMAIL THIS PAGE TO A FRIEND
03696 Sigma-Aldrich

Fmoc-β-Homoser(tBu)-OH

≥97.5% (HPLC)

Synonym: Fmoc-O-tert-butyl-L-β-homoserine

  • CAS Number 203854-51-7

  • Empirical Formula (Hill Notation) C23H27NO5

  • Molecular Weight 397.46

  •  Beilstein Registry Number 8014825

  •  MDL number MFCD01862865

  •  PubChem Substance ID 24845558

Purchase

Properties

Related Categories Beta-Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   SKYFRYZPXHNPOK-OAHLLOKOSA-N
assay   ≥97.5% (HPLC)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

β-Amino Acids and Homologs

Although they are less abundant than their α-analogues, β-amino acids occur in nature both in free form and bound to peptides. Oligomers composed exclusively of β-amino acids (so-called β-peptides) m...
Matthias Junkers
Aldrich ChemFiles 2008, 8.7, 11.
Keywords: Chemfiles, Degradations, Metabolism, Peptidomimetics

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?