• USA Home
  • 03697 - Boc-β-Homoser(Bzl)-OH

EMAIL THIS PAGE TO A FRIEND
03697 Sigma-Aldrich

Boc-β-Homoser(Bzl)-OH

≥98.0% (HPLC)

Synonym: Boc-O-benzyl-L-β-homoserine

  • CAS Number 218943-31-8

  • Empirical Formula (Hill Notation) C16H23NO5

  • Molecular Weight 309.36

  •  Beilstein Registry Number 8070512

  •  MDL number MFCD01862866

  •  PubChem Substance ID 24845559

Purchase

Properties

Related Categories Beta-Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   BNSXKDBZQDOLAL-CYBMUJFWSA-N
assay   ≥98.0% (HPLC)
optical activity   [α]20/D +22.0±1.0°, c = 1 in methanol

Description

Other Notes

Building block for the synthesis of L-β-homoserine-derived β2,3 -amino acids by β-alkylation and peptides containing these amino acids

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

β-Amino Acids and Homologs

Although they are less abundant than their α-analogues, β-amino acids occur in nature both in free form and bound to peptides. Oligomers composed exclusively of β-amino acids (so-called β-peptides) m...
Matthias Junkers
Aldrich ChemFiles 2008, 8.7, 11.
Keywords: Chemfiles, Degradations, Metabolism, Peptidomimetics

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?