• USA Home
  • 03700 - Ethylenediaminetetraacetic acid tetrasodium salt hydrate

EMAIL THIS PAGE TO A FRIEND
03700 Sigma-Aldrich

Ethylenediaminetetraacetic acid tetrasodium salt hydrate

≥98.0% (calc. based on dry substance, KT)

Synonym: Tetrasodium ethylenediaminetetraacetate hydrate

Purchase

Properties

InChI Key   GXFFSKMBMYHTAE-UHFFFAOYSA-J
assay   ≥98.0% (calc. based on dry substance, KT)
impurities   ~7.5% water
mp   >300 °C(lit.)
solubility   H2O: 0.1 g/mL clear, colorless

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Respiratory Tract
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?