EMAIL THIS PAGE TO A FRIEND
03735 Sigma-Aldrich

Benzo-1,4-dioxane

purum, ≥98.0%

Synonym: 1,2-Ethylenedioxybenzene, 1,4-Benzodioxan, 2,3-Dihydro-1,4-benzodioxin

  • CAS Number 493-09-4

  • Empirical Formula (Hill Notation) C8H8O2

  • Molecular Weight 136.15

  •  Beilstein Registry Number 120846

  •  EC Number 207-775-6

  •  MDL number MFCD00006821

  •  PubChem Substance ID 329747788

Purchase

Properties

InChI Key   BNBQRQQYDMDJAH-UHFFFAOYSA-N
grade   purum
assay   ≥98.0%
refractive index   n20/D 1.549(lit.)
  n20/D 1.550
bp   103 °C/6 mmHg(lit.)
density   1.142 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
DF4728000
Flash Point(F) 
156 °F
Flash Point(C) 
69 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?