• USA Home
  • 03754 - Boc-β-(3-thienyl)-D-Ala-OH (dicyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
03754 Sigma-Aldrich

Boc-β-(3-thienyl)-D-Ala-OH (dicyclohexylammonium) salt

≥98.0% (TLC)

Synonym: Boc-3-(3-thienyl)-D-alanine (dicyclohexylammonium) salt

  • CAS Number 331730-14-4

  • Empirical Formula (Hill Notation) C12H17NO4S · C12H23N

  • Molecular Weight 452.65

  •  MDL number MFCD01860888

  •  PubChem Substance ID 329747795

Purchase

Properties

Related Categories Alanine Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   FNTVEJPWKBETJW-SBSPUUFOSA-N
assay   ≥98.0% (TLC)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?