• USA Home
  • 03793 - Silicon atomic spectroscopy standard concentrate 1.00 g Si

EMAIL THIS PAGE TO A FRIEND
03793 Sigma-Aldrich

Silicon atomic spectroscopy standard concentrate 1.00 g Si

1.00 g/L, for 1L standard solution, analytical standard

Synonym: Sodium silicate solution, Sodium trisilicate solution, Water glass

  • Linear Formula Na2O(SiO2)x · xH2O

Purchase

Properties

Related Categories 1.00 g/L, AAS, AAS Concentrates, Additional Standards, Analytical Standards,
grade   analytical standard
shelf life   limited shelf life, expiry date on the label
packaging   ampule of
solubility   H2O: soluble

Description

Preparation Note

prepared with sodium silicate and H2O

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 1824 8 / PGII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?