• USA Home
  • 03794 - Strontium atomic spectroscopy standard concentrate 1.00 g Sr

EMAIL THIS PAGE TO A FRIEND
03794 Sigma-Aldrich

Strontium atomic spectroscopy standard concentrate 1.00 g Sr

1.00 g/L, for 1L standard solution, analytical standard

Synonym: Strontium chloride solution

Purchase

Properties

Related Categories AAS, Additional Standards, Analytical Standards, Analytical/Chromatography, Elemental & Metal Standards,
grade   analytical standard
InChI Key   CIOAGBVUUVVLOB-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
packaging   ampule of
solubility   H2O: soluble

Description

Preparation Note

prepared with SrCl2 and H2O

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?