• USA Home
  • 03797 - Vanadium atomic spectroscopy standard concentrate 1.00 g V

EMAIL THIS PAGE TO A FRIEND
03797 Sigma-Aldrich

Vanadium atomic spectroscopy standard concentrate 1.00 g V

1.00 g/L, for 1L standard solution, analytical standard

Synonym: Ammonium (meta)vanadate solution

Purchase

Properties

Related Categories 1.00 g/L, AAS, AAS Concentrates, Additional Standards, Analytical Standards,
grade   analytical standard
InChI Key   LEONUFNNVUYDNQ-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
packaging   ampule of
solubility   H2O: soluble

Description

Preparation Note

prepared with NH4VO3, NaOH and H2O

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN3289 - class 6.1 - PG 2 - basic - Toxic liquid, corrosive, inorganic, n.o.s., HI: all
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?