• USA Home
  • 03867 - Turkey red oil sodium salt

EMAIL THIS PAGE TO A FRIEND
03867 Sigma-Aldrich

Turkey red oil sodium salt

100%

Synonym: Castor-oil sulfated sodium salt, Sulfated castor oil, Sulforicinolate sodium salt

Purchase

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
228.2 °F
Flash Point(C) 
109 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

16316 Turkey red oil sodium salt, for microscopy

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?