EMAIL THIS PAGE TO A FRIEND
03892 Sigma-Aldrich

Hayem’s solution

for counting (of erythrocytes)

Purchase

Properties

grade   for counting (of erythrocytes)
impurities   mercuric chloride

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Hazard statements 
Precautionary statements 
RIDADR 
UN 2024 6.1 / PGII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?