• USA Home
  • 03893 - Nylander’s solution

EMAIL THIS PAGE TO A FRIEND
03893 Sigma-Aldrich

Nylander’s solution

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Gravimetric Reagents More...
impurities   sodium hydroxide
density   1.14 g/mL at 20 °C

Description

Application

reagent for determination of glucose

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1824 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?