• USA Home
  • 03973 - Hematoxylin solution A according to Weigert

EMAIL THIS PAGE TO A FRIEND
03973 Sigma-Aldrich

Hematoxylin solution A according to Weigert

for microscopy

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Microscopy Reagents, Stains and Dyes
grade   for microscopy
InChI Key   WZUVPPKBWHMQCE-XJKSGUPXSA-N
concentration   100 g/L
impurities   ethanol
density   0.84 g/mL at 20 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1170 - class 3 - PG 2 - Ethanol, solution
WGK Germany 
3
Flash Point(F) 
57.2 °F
Flash Point(C) 
14 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?