• USA Home
  • 03974 - Hematoxylin solution B according to Weigert

EMAIL THIS PAGE TO A FRIEND
03974 Sigma-Aldrich

Hematoxylin solution B according to Weigert

for microscopy

Synonym: Lithium carbonate solution

Purchase

Properties

grade   for microscopy
InChI Key   XGZVUEUWXADBQD-UHFFFAOYSA-L
impurities   lithium carbonate
density   1.00 g/mL at 20 °C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?