• USA Home
  • 04026 - Rhodium ICP-MS Standard

EMAIL THIS PAGE TO A FRIEND
04026 Sigma-Aldrich

Rhodium ICP-MS Standard

ready-to-use, 10 mg/kg Rh in nitric acid

Synonym: Rhodium(III) nitrate solution

  • Linear Formula Rh(NO3)3

  • Molecular Weight 288.92

  •  MDL number MFCD00011201

  •  PubChem Substance ID 24845638

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Elemental & Metal Standards, Elemental, Ions and Water (Karl Fischer) Standards,
InChI Key   MHOVAHRLVXNVSD-UHFFFAOYSA-N
CofA   the measured values of the metal traces are reported on Certificate of Analysis
shelf life   limited shelf life, expiry date on the label
concentration   10 mg/kg Rh in nitric acid

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Preparation Note

prepared with Rh(NO3)3 and HNO3 (∼0.5 M)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?