EMAIL THIS PAGE TO A FRIEND
04080 Sigma-Aldrich

2-Ethyl-1-hexylamine

purum, ≥98.0% (GC)

  • CAS Number 104-75-6

  • Linear Formula CH3(CH2)3CH(C2H5)CH2NH2

  • Molecular Weight 129.24

  •  Beilstein Registry Number 1209249

  •  EC Number 203-233-8

  •  MDL number MFCD00008148

  •  PubChem Substance ID 329747876

Purchase

Properties

vapor pressure   1.2 mmHg ( 20 °C)
InChI Key   LTHNHFOGQMKPOV-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
autoignition temp.   563 °F
refractive index   n20/D 1.431(lit.)
  n20/D 1.431
bp   169 °C(lit.)
mp   −76 °C(lit.)
density   0.789 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2276 8(3) / PGIII
WGK Germany 
2
RTECS 
MQ5250000
Flash Point(F) 
122 °F
Flash Point(C) 
50 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

E29508 2-Ethyl-1-hexylamine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?