• USA Home
  • 04130 - N-Ethyl-N-(2-hydroxyethyl)-p-phenylenediamine sulfate salt monohydrate

EMAIL THIS PAGE TO A FRIEND
04130 Sigma-Aldrich

N-Ethyl-N-(2-hydroxyethyl)-p-phenylenediamine sulfate salt monohydrate

≥97.0% (T)

  • CAS Number 4327-84-8

  • Empirical Formula (Hill Notation) C10H16N2O · H2SO4 · H2O

  • Molecular Weight 296.34

  •  Beilstein Registry Number 3789014

  •  EC Number 224-361-0

  •  MDL number MFCD00150603

  •  PubChem Substance ID 329747886

Purchase

Properties

Related Categories Amino Alcohols, Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds More...
InChI Key   HUISMDQQFGXRKT-UHFFFAOYSA-N
assay   ≥97.0% (T)
mp   177-179 °C (dec.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?