EMAIL THIS PAGE TO A FRIEND
04220 Sigma-Aldrich

2-Methyl-3-pentanone

purum, ≥99.0% (GC)

Synonym: Ethyl isopropyl ketone

  • CAS Number 565-69-5

  • Linear Formula (CH3)2CHCOC2H5

  • Molecular Weight 100.16

  •  Beilstein Registry Number 1698849

  •  EC Number 209-288-4

  •  MDL number MFCD00009314

  •  PubChem Substance ID 329747894

Purchase

Properties

Related Categories Building Blocks, C3 to C6, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   HYTRYEXINDDXJK-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.397(lit.)
  n20/D 1.398
bp   113 °C(lit.)
density   0.811 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1224 - class 3 - PG 2 - VP > 110kPa - Ketones, liquid, n.o.s., HI: all
WGK Germany 
3
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?