04415 Sigma-Aldrich

Sodium hypophosphite monohydrate

puriss., meets analytical specification of DAC, 99-103%

DOWNLOAD MSDS (PDF)
  • CAS Number 10039-56-2

  • Linear Formula NaH2PO2 · H2O

  • Molecular Weight 105.99

  •  EC Number 231-669-9

  •  MDL number MFCD06252379

  •  PubChem Substance ID 24845737

Properties

Related Categories Chemical Synthesis, Inorganic Salts, Sodium, Synthetic Reagents
grade   puriss.
assay   99-103%
quality   meets analytical specification of DAC
impurities   ≤0.001% heavy metals (as Pb)
anion traces   chloride (Cl-): ≤200 mg/kg
  sulfate (SO42-): ≤200 mg/kg
cation traces   As: ≤5 mg/kg
  Ca: ≤100 mg/kg
  Fe: ≤10 mg/kg

Price and Availability

Customers Also Viewed

Sodium hypophosphite

puriss., meets analytical specification of BPC63, anhydrous, 98-101%

Sodium hypophosphite monohydrate

puriss. p.a., Reag. Ph. Eur., ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?