EMAIL THIS PAGE TO A FRIEND
04501 Sigma-Aldrich

Calcium phosphide

≥8% active phoshor (P) basis

Purchase

Properties

Related Categories Calcium, Chemical Synthesis, Inorganic Salts, Synthetic Reagents
InChI Key   HMTIWQQJRUFNAF-UHFFFAOYSA-N
assay   ≥8% active phoshor (P) basis

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Contact with water liberates toxic gas.
RIDADR 
UN1360 - class 4.3 - PG 1 - Calcium phosphide
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?