EMAIL THIS PAGE TO A FRIEND
04800 Sigma-Aldrich

4-Ethylpyridine

purum, ≥97.0% (GC)

  • CAS Number 536-75-4

  • Empirical Formula (Hill Notation) C7H9N

  • Molecular Weight 107.15

  •  Beilstein Registry Number 106481

  •  EC Number 208-646-7

  •  MDL number MFCD00006450

  •  PubChem Substance ID 329747981

Purchase

Properties

InChI Key   VJXRKZJMGVSXPX-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.501(lit.)
  n20/D 1.502
bp   168 °C(lit.)
density   0.942 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
122 °F
Flash Point(C) 
50 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

112437 4-Ethylpyridine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?