EMAIL THIS PAGE TO A FRIEND
04890 Sigma-Aldrich

N-Ethylthiourea

purum, ≥98.0% (N)

Synonym: 1-Ethylthiourea

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thioureas More...
InChI Key   GMEHFXXZSWDEDB-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (N)
mp   108-110 °C(lit.)
  108-110 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
YT3700000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

252530 N-Ethylthiourea, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?