• USA Home
  • 05032 - Aflatoxin B1 from Aspergillus flavus

EMAIL THIS PAGE TO A FRIEND
05032 Sigma-Aldrich

Aflatoxin B1 from Aspergillus flavus

from Aspergillus flavus

  • CAS Number 1162-65-8

  • Empirical Formula (Hill Notation) C17H12O6

  • Molecular Weight 312.27

  •  Beilstein Registry Number 1269174

  •  EC Number 214-603-3

  •  MDL number MFCD00869647

  •  PubChem Substance ID 329748011

Purchase

Properties

biological source   from Aspergillus flavus
InChI Key   OQIQSTLJSLGHID-WNWIJWBNSA-N
storage temp.   2-8°C

Description

Warning

Possibly carcinogenic.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3462 6.1 / PGI
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A6636 Aflatoxin B1 from Aspergillus flavus, from Aspergillus flavus

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?